Learn More
CD11b/c Mouse anti-Rat, FITC, Clone: OX-42, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561691
Additional Details : Weight : 0.01000kg
Description
The OX-42 monoclonal antibody specifically binds to the CR3 complement (C3bi) receptor found on most monocytes, granulocytes, macrophages, dendritic cells, and microglia. It appears to recognize a common epitope shared by CD11b and CD11c (integrin αM and αX chains). OX-42 antibody inhibits C3bi binding activity.
Specifications
CD11b/c | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Resident peritoneal cells | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
OX-42 | |
FITC | |
Mouse | |
Affinity Purified | |
RUO | |
Rat | |
IgG2a κ |
For Research Use Only.