missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11b Antibody (M1/70.15), PE/Cy5.5, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB600-1327PECY55
This item is not returnable.
View return policy
Description
CD11b Monoclonal specifically detects CD11b in Human, Mouse, Rabbit samples. It is validated for Flow Cytometry.
Specifications
| CD11b | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| ITGAM | |
| Rat Monoclonal CD11b Antibody (M1/70.15) was made to T cell enriched splenocytes from B10 mice. | |
| Protein G purified | |
| Breast Cancer, Cancer, Cellular Markers, Cytoskeleton Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 3684.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| M1/70.15 | |
| Flow Cytometry | |
| antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement | |
| Rat | |
| 127.2 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rabbit | |
| IgG2b λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction