missing translation for 'onlineSavingsMsg'
Learn More

CD11a/CD18 Antibody (hu1124 (Efalizumab)), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18376401 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376401 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376401 Supplier Novus Biologicals Supplier No. NBP275912AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

CD11a/CD18 Monoclonal specifically detects CD11a/CD18 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11a/CD18
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone hu1124 (Efalizumab)
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1
Gene Symbols ITGAL
Host Species Human
Immunogen Efalizumab binds to CD11a, a subunit of leukocyte function antigen-1 (LFA-1), which is expressed on all leukocytes.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3683
Test Specificity Binds to human LFA-1.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.