Learn More
CD119 (IFN gamma Receptor 1) Monoclonal Antibody (2E2), PE, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Affymetrix eBioscience 12-1191-80
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal antibody 2E2 recognizes mouse CD119 also known as IFN gamma receptor1. CD119 heterodimerizes with IFN gamma receptor 2 (AF-1) forming the Class II cytokine receptor. The ligand IFN gamma binds IFN gamma receptor 1 subunit but signal transduction transpires via the IFN gamma receptor 2 subunit. The receptor is involved in host defense and expression is broad including B cells, NK cells, macrophages and dendritic cells as well as other non-hematopoietic cell types. Applications Reported: This 2E2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2E2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. In addition, longer incubations appear to help stain the popµLation; therefore we recommend a 60 minute incubation at 2-8°C. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
This gene (IFNGR1) encodes the ligand-binding chain (alpha) of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. A genetic variation in IFNGR1 is associated with susceptibility to Helicobacter pylori infection. In addition, defects in IFNGR1 are a cause of mendelian susceptibility to mycobacterial disease, also known as familial disseminated atypical mycobacterial infection.Specifications
CD119 (IFN gamma Receptor 1) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P15261 | |
IFNGR1 | |
Affinity chromatography | |
RUO | |
15979 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
2E2 | |
PE | |
IFNGR1 | |
antiviral protein, type 2; AVP, type 2; CD119; CD119 antigen; CDw119; Ifgr; IFN-gamma receptor 1; IFN-gammaR; IFN-gamma-R1; IFN-gamma-R-alpha; Ifngr; IFNGR1; IMD27A; IMD27B; immune interferon receptor 1; INF-g receptor; interferon gamma receptor 1; Interferon gamma receptor alpha-chain; interferon-gamma receptor 1; interferon-gamma receptor alpha chain; Nktar | |
Armenian Hamster | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
For Research Use Only.