Learn More
CD117 Mouse anti-Human, PE, Clone: YB5.B8, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555714
Description
The YB5.B8 monoclonal antibody specifically binds to CD117. CD117, also known as c-Kit, is a 145 kDa cell-surface glycoprotein with tyrosine kinase activity. CD117 is present on hematopoietic progenitor cell subsets, thymocytes, mast cells, hepatocytes and histiocytes. CD117 serves as a cytokine receptor for steel factor (SLF), also known as stem cell factor (SCF) or mast cell growth factor (MGF). The interaction of c-Kit and SLF is crucial to hematopoiesis, mast cell differentiation, melanogenesis, and germ cell development. The ability of the YB5.B8 antibody to block the binding of c-Kit ligand is still controversial.
Specifications
| CD117 | |
| Monoclonal | |
| 0.2mg/mL | |
| KIT; c-Kit; SCFR; PBT; Mast/stem cell growth factor receptor | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| YB5.B8 | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Acute myelocytic leukemia blasts | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.