missing translation for 'onlineSavingsMsg'
Learn More

CD117 Mouse anti-Human, PE, Clone: YB5.B8, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 561682

 View more versions of this product

Product Code. 15871229

  • ONLINE EXCLUSIVE PRICE
    £124.00
    £117.51 / Each
    Save £6.49
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The YB5.B8 monoclonal antibody specifically binds to CD117. CD117, also known as c-Kit, is a 145 kDa cell-surface glycoprotein with tyrosine kinase activity. CD117 is present on hematopoietic progenitor cell subsets, thymocytes, mast cells, hepatocytes and histiocytes. CD117 serves as a cytokine receptor for steel factor (SLF), also known as stem cell factor (SCF) or mast cell growth factor (MGF). The interaction of c-Kit and SLF is crucial to hematopoiesis, mast cell differentiation, melanogenesis, and germ cell development. The ability of the YB5.B8 antibody to block the binding of c-Kit ligand is still controversial.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD117
Monoclonal
0.2mg/mL
KIT; c-Kit; SCFR; PBT; Mast/stem cell growth factor receptor
Mouse
Affinity Purified
RUO
Human
IgG1 κ
Flow Cytometry
YB5.B8
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Acute myelocytic leukemia blasts
25 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.