Learn More
CD117 (c-Kit) Rat, anti-Mouse, Unconjugated, Clone: ACK2, STEMCELL Technologies
Rat Monoclonal Antibody
Brand: Stemcell Technologies 60034.1
This item is not returnable.
View return policy
Description
- The ACK2 antibody reacts with CD117 (c-Kit), an ∼145 kDa type 1 transmembrane receptor for c-Kit ligand (stem cell factor/steel factor) that is broadly expressed on hematopoietic stem cells in bone marrow, including pluripotent and erythroid progenitor cells and B and T lymphocyte precursors, as well as on mast cells
- CD117 belongs to the tyrosine kinase receptor family and possesses five immunoglobulin-like C2-type domains and a cytoplasmic protein kinase domain
- Binding of c-Kit ligand to CD117 induces dimerization and autophosphorylation, which activates several intracellular signaling pathways critical for the proliferation and differentiation of hematopoietic stem cells
- Signaling by CD117 is modulated by phosphatases and by rapid endocytosis and degradation of the receptor
- Mutations in CD117 are associated with various types of tumors and the piebald trait, an autosomal dominant abnormality of pigmentation
Specifications
| CD117 (c-Kit) | |
| Monoclonal | |
| Unconjugated | |
| 50 μg | |
| Mouse | |
| Hematopoietic Stem and Progenitor Cells |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| ACK2 | |
| Rat | |
| Primary | |
| IgG2b κ |