missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117 (c-Kit) Rat, anti-Mouse, APC, Clone: ACK2, STEMCELL Technologies
Rat Monoclonal Antibody
£153.90 - £355.05
Specifications
| Antigen | CD117 (c-Kit) |
|---|---|
| Clone | ACK2 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17178661
|
Stemcell Technologies
60034AZ.1 |
25 μg |
£153.90
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17168661
|
Stemcell Technologies
60034AZ |
100 μg |
£355.05
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The ACK2 antibody reacts with CD117 (c-Kit), an ∼145 kDa type 1 transmembrane receptor for c-Kit ligand (stem cell factor/steel factor) that is broadly expressed on hematopoietic stem cells in bone marrow, including pluripotent and erythroid progenitor cells and B and T lymphocyte precursors, as well as on mast cells
- CD117 belongs to the tyrosine kinase receptor family and possesses five immunoglobulin-like C2-type domains and a cytoplasmic protein kinase domain
- Binding of c-Kit ligand to CD117 induces dimerization and autophosphorylation, which activates several intracellular signaling pathways critical for the proliferation and differentiation of hematopoietic stem cells
- Signaling by CD117 is modulated by phosphatases and by rapid endocytosis and degradation of the receptor
- Mutations in CD117 are associated with various types of tumors and the piebald trait, an autosomal dominant abnormality of pigmentation
Specifications
| CD117 (c-Kit) | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| APC | |
| Mouse | |
| Primary |
| ACK2 | |
| Monoclonal | |
| Rat | |
| IgG2b κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title