Learn More
Invitrogen™ CD117 (c-Kit) Monoclonal Antibody (2B8), FITC, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£79.50 - £238.00
Specifications
| Antigen | CD117 (c-Kit) |
|---|---|
| Clone | 2B8 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15268169
|
Invitrogen™
11-1171-81 |
50 μg |
£79.50
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15278169
![]() |
Invitrogen™
11-1171-82 |
100 μg |
£146.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15288169
|
Invitrogen™
11-1171-85 |
500 μg |
£238.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The 2B8 monoclonal antibody reacts with mouse CD117, also known as c-Kit receptor, Steel factor receptor, and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145-kDa molecule is expressed by a majority of hematopoietic progenitor cells characterized in the mouse bone marrow as a small subset of cells positive for Sca-1 and Thy1 (Thy1 low) and negative for lineage markers. The interaction of the mouse c-Kit receptor and steel factor promotes the proliferation and differentiation of hematopoietic progenitor cells. CD117 is also expressed by mast cells and plays a role in signaling and activation of these cells. Applications Reported: The 2B8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2B8 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
KIT (c-KIT) is a proto-oncogene and a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). KIT was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit. KIT together with its ligand regulates growth and activation of a variety of hemopoietic and non-hemopoietic cells. Mutations in KIT are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous leukemia, and piebaldism. Recently, deregulation of the KIT receptor TK by the prevalent activation loop mutation D816V has served as a focal point in therapeutic strategies aimed at curbing neoplastic mast cell growth. c-Kit is expressed in hematopoietic stem cells, germ cells, mast cells and gastrointestinal tract cajal cells. Upon binding of its ligand stem cell factor (SCF), c-kit dimerizes, resulting in receptor activation and autophosphorylation of various tyrosine residues including tyrosine 703 located on the cytoplasmic domain of the receptor. This modification allows docking of Grb2 and activation of the Ras/ERK signaling pathway. SCF/c-kit can activate multiple downstream signaling pathways including PI3K, PLC-gamma and JAK/STAT. c-kit receptor activation is essential for hematopoiesis, stem cell maintenance and gametogenesis.Specifications
| CD117 (c-Kit) | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| belly-spot; Bs; CD117; ckit; C-Kit; c-KIT gene1; c-kit protein; c-kit proto-oncogene protein; c-kit receptor; c-kit receptor tyrosine kinase; dominant spotting; Dominant white spotting; Fdc; Gsfsco1; Gsfsco5; Gsfsow3; KIT; kit oncogene; KIT proto-oncogene receptor tyrosine kinase; KIT proto-oncogene, receptor tyrosine kinase; mast cell growth factor receptor; mast/stem cell growth factor receptor; mast/stem cell growth factor receptor Kit; MGF; p145 c-kit; PBT; Piebald trait protein; protein kinase; proto-oncogene c-Kit; proto-oncogene tyrosine-protein kinase Kit; receptor tyrosine kinase c-kit; receptor tyrosine kinase type III; SCFR; SCO1; SCO5; Sl; soluble KIT variant 1; SOW3; spotted sterile male; Ssm; Steel Factor Receptor; Tr-kit; tyrosine kinase receptor; tyrosine kinase receptor c-KIT; tyrosine-protein kinase Kit; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein; W | |
| KIT | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| KIT |
| 2B8 | |
| Flow Cytometry | |
| FITC | |
| Rat | |
| Mouse, Pig | |
| P05532, Q2HWD6 | |
| 16590, 396810 | |
| IgG2b κ | |
| Affinity chromatography | |
| Antibody |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.