missing translation for 'onlineSavingsMsg'
Learn More

CD117/c-kit Antibody (ACK2), DyLight 680, Novus Biologicals™

Product Code. 18404014 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404014 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404014 Supplier Novus Biologicals Supplier No. NBP143359FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

CD117/c-kit Monoclonal antibody specifically detects CD117/c-kit in Human, Mouse, Guinea Pig samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunohistochemistry (Whole-Mount)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD117/c-kit
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone ACK2
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein
Host Species Rat
Immunogen Mouse CD117
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3815
Target Species Human, Mouse, Guinea Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.