missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD115 (CSF-1R) Rat anti-Mouse , PE, Clone: AFS98 (also known as AFS98), BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 566839
This item is not returnable.
View return policy
Description
CD115 (CSF-1R) Monoclonal antibody specifically detects CD115 (CSF-1R) in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD115 (CSF-1R) | |
| Monoclonal | |
| PE | |
| CD115; CSF-1R; Csf1r; M-CSFR; Fms; c-fms | |
| Mouse | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| AFS98 (also known as AFS98) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction