missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD115 (c-fms) Rat anti-Mouse, PE, Clone: AFS98, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech PE-65095-100UG
This item is not returnable.
View return policy
Description
The CD115 (c-fms) antibody from Proteintech is a rat monoclonal antibody to a n/a of mouse CD115 (c-fms). This antibody recognizes mouse antigen. The CD115 (c-fms) antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifications
| CD115 (c-fms) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| Primary | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| AFS98 | |
| PE | |
| 100 μg | |
| 12978 | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction