missing translation for 'onlineSavingsMsg'
Learn More

CD115 (c-fms) Rat anti-Mouse, APC, Clone: AFS98, Proteintech

Product Code. 17254656 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17254656 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 17254656 Supplier Proteintech Supplier No. APC65095100UG
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Rat Monoclonal Antibody

The CD115 (c-fms) antibody from Proteintech is a rat monoclonal antibody to a n/a of mouse CD115 (c-fms). This antibody recognizes mouse antigen. The CD115 (c-fms) antibody has been validated for the following applications: FC analysis. This antibody is apc.

Specifications

Antigen CD115 (c-fms)
Applications Flow Cytometry
Classification Monoclonal
Clone AFS98
Concentration 0.2 mg/mL
Conjugate APC
Host Species Rat
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 12978
Target Species Mouse
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.