missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD115 (c-fms) Monoclonal Antibody (AFS98), NovaFluor™ Red 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£154.00 - £297.00
Specifications
| Antigen | CD115 (c-fms) |
|---|---|
| Clone | AFS98 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30054803
|
Invitrogen™
M041T03R04-A |
100 μg |
£297.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30056047
|
Invitrogen™
M041T02R04-A |
25 μg |
£154.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD115 (c-fms) Monoclonal antibody specifically detects CD115 (c-fms) in Mouse samples. It is validated for Flow CytometrySpecifications
| CD115 (c-fms) | |
| 0.1 mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| P09581 | |
| 12978 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| AFS98 | |
| Flow Cytometry | |
| NovaFluor Red 710 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| AI323359; CD115; CD115 antigen; C-FMS; colony stimulating factor 1 receptor; colony stimulating factor 1 receptor, formerly McDonough feline sarcoma viral (v-fms) oncogene homolog; CSF-1 receptor; CSF1R; CSF-1R; CSF-1-R; CSFIR; Csfmr; CSFR; EC 2.7.10.1; FIM2; Fim-2; FMS; fms proto-oncogene; HDLS; kinase CSFR; macrophage colony stimulating factor I receptor; macrophage colony-stimulating factor 1 receptor; McDonough feline sarcoma viral (v-fms) oncogene homolog; M-CSFR; M-CSF-R; mrfms; OTTHUMP00000224093; protein-tyrosine kinase; proto-oncogene c-Fms; proto-oncogene fms | |
| IgG2a κ | |
| Affinity chromatography | |
| Csf1r |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title