missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD109 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£350.00 - £507.00
Specifications
| Antigen | CD109 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18375012
|
Novus Biologicals
NBP3-17814-25UL |
25 μg |
£350.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18301894
|
Novus Biologicals
NBP3-17814-100UL |
100 μg |
£507.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD109 Polyclonal antibody specifically detects CD109 in Human samples. It is validated for ImmunofluorescenceSpecifications
| CD109 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers | |
| PBS, pH 7.2, 40% glycerol | |
| 135228 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 150 kDa TGF-beta-1-binding protein, activated T-cell marker CD109, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7, CD109 antigen, CD109 antigen (Gov platelet alloantigens), CD109 molecule, CPAMD7r150, DKFZp762L1111, FLJ38569, FLJ41966, Gov platelet alloantigens, p180, Platelet-specific Gov antigen, RP11-525G3.1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: VVSGNKRLKELSYMVVSRGQLVAVGKQNSTMFSLTPENSWTPKACVIVYYIEDDGEIISDVLKIPVQLVFKNKIKLYWSKVKAEPSEKVSLRISVTQPDSIVGIVAVDKSVNLMNASNDITMENVVHEL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title