missing translation for 'onlineSavingsMsg'
Learn More

CD109 Antibody (W7C5), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18430837 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430837 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430837 Supplier Novus Biologicals Supplier No. NBP275969MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD109 Monoclonal antibody specifically detects CD109 in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD109
Applications Flow Cytometry
Classification Monoclonal
Clone W7C5
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Flow (Cell Surface)
Formulation 50mM Sodium Borate
Gene Alias 150 kDa TGF-beta-1-binding protein, activated T-cell marker CD109, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7, CD109 antigen, CD109 antigen (Gov platelet alloantigens), CD109 molecule, CPAMD7r150, DKFZp762L1111, FLJ38569, FLJ41966, Gov platelet alloantigens, p180, Platelet-specific Gov antigen, RP11-525G3.1
Host Species Mouse
Immunogen WERI-RB-1 retinoblastoma cell line
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 135228
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.