missing translation for 'onlineSavingsMsg'
Learn More

CD109 Antibody [DyLight 755], Novus Biologicals™

Product Code. 30733920 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733920 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733920 Supplier Novus Biologicals Supplier No. AF7717Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

CD109 Polyclonal antibody specifically detects CD109 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD109
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias 150 kDa TGF-beta-1-binding protein, activated T-cell marker CD109, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7, CD109 antigen, CD109 antigen (Gov platelet alloantigens), CD109 molecule, CPAMD7r150, DKFZp762L1111, FLJ38569, FLJ41966, Gov platelet alloantigens, p180, Platelet-specific Gov antigen, RP11-525G3.1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse CD109, Ala22-Ser1269, Accession # Q8R422
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 135228
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.