missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD109 Antibody (B-E47), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18101CL1
This item is not returnable.
View return policy
Description
CD109 Monoclonal antibody specifically detects CD109 in Human samples. It is validated for Flow Cytometry
Specifications
| CD109 | |
| Monoclonal | |
| CoraFluor 1 | |
| 150 kDa TGF-beta-1-binding protein, activated T-cell marker CD109, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7, CD109 antigen, CD109 antigen (Gov platelet alloantigens), CD109 molecule, CPAMD7r150, DKFZp762L1111, FLJ38569, FLJ41966, Gov platelet alloantigens, p180, Platelet-specific Gov antigen, RP11-525G3.1 | |
| Eahy 926 cell line. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells | |
| 0.1 mL | |
| Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers | |
| 135228 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry | |
| B-E47 | |
| PBS | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction