missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD108 Mouse anti-Human, BUV615, Clone: KS-2, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 751594
This item is not returnable.
View return policy
Description
CD108 Monoclonal antibody specifically detects CD108 in Human samples. It is validated for Flow Cytometry
Specifications
| CD108 | |
| Monoclonal | |
| Brilliant Ultraviolet 615 | |
| JMH; SEM7A; Sema K1; Sema L; John-Milton-Hargen human blood group Ag | |
| BALB/c splenocytes immunized with PHA-activated human PBMCs fused with murine cell line NS-1 | |
| RUO | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| KS-2 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction