missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD107a (LAMP-1) Monoclonal Antibody (eBioH4A3), Brilliant Violet™ 711, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£153.00 - £321.00
Specifications
| Antigen | CD107a (LAMP-1) |
|---|---|
| Clone | eBioH4A3 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30245350
![]() |
Invitrogen™
407-1079-42 |
100 Tests |
£321.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30245314
|
Invitrogen™
407-1079-41 |
25 Tests |
£153.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD107a (LAMP-1) Monoclonal antibody specifically detects CD107a (LAMP-1) in Human samples. It is validated for Flow CytometrySpecifications
| CD107a (LAMP-1) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 120 kDa lysosomal membrane glycoprotein; AI196048; CD107 antigen-like family member A; CD107a; I79_011073; Lamp I; LAMP1; LAMP-1; LAMPA; LGP120; LGP-120; LGPA; LGP-A; lysosomal associated membrane protein 1; Lysosomal associated membrane protein 1 (120 kDa); lysosomal membrane glycoprotein 1; lysosomal membrane glycoprotein A; lysosomal-associated membrane protein 1; lysosome-associated membrane glycoprotein 1; LYSOSOME-ASSOCIATED MEMBRANE GLYCOPROTEIN 1 PRECURSOR (LAMP-1) (LGP-A) (LGP-120) (CD107A) (P2B); lysosome-associated membrane protein 1; P2B | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| eBioH4A3 | |
| Flow Cytometry | |
| Brilliant Violet 711 | |
| Mouse | |
| Human | |
| P11279 | |
| 3916 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| LAMP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title