Learn More
CD106 (VCAM-1) Monoclonal Antibody (eBioMR106 (MR106)), Biotin, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 13-1060-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The eBioMR106 monoclonal antibody reacts with rat CD106 (Vascular Cell Adhesion Molecule-1, VCAM-1), a 110 kDa transmembrane glycoprotein expressed by myeloid lineage and bone marrow stromal cells. Endothelial cells constitutively express low levels of CD106 and further upregulate VCAM-1 upon cytokine stimulation. CD106 binds to integrin alpha4beta1 (VLA-4, CD49d/CD29) and Integrin alpha4beta7 (LPAM-1) and these interactions in the bone marrow and thymus are important for early lymphocyte and myeloid development. Cytokine-mediated upregulation of CD106 on endothelial cells suggests a role for this antigen in the inflammatory response. Applications Reported: This eBioMR106 (MR106) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioMR106 (MR106) antibody has been tested by flow cytometric analysis of rat bone marrow cells. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.Specifications
CD106 (VCAM-1) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P29534 | |
VCAM1 | |
Affinity chromatography | |
RUO | |
25361 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
eBioMR106 (MR106) | |
Biotin | |
VCAM1 | |
adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B | |
Mouse | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 κ |
For Research Use Only.