Learn More
CD105 Monoclonal Antibody (SN6), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD10505
Code : 50
Additional Details : Weight : 0.50000kg
Description
Allophycocyanin (APC) is a stable and highly soluble phycobiliprotein that provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing exceptional quantum yields and molar extinction coefficients.
CD105 Monoclonal antibody specifically detects CD105 in Human samples. It is validated for Flow Cytometry.Specifications
CD105 | |
Monoclonal | |
APC | |
ENG | |
AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein | |
Mouse | |
purified | |
ASR, ASR/IVD | |
2022 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
SN6 | |
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide | |
P17813 | |
ENG | |
Human CD105 (Endoglin) | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |