missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD105 (Endoglin) Mouse, anti-Human, Biotin, Clone: 43A3, STEMCELL Technologies
Description
- The 43A3 antibody reacts with CD105 (endoglin), an ∼180 kDa cell surface glycoprotein which is a disulfide-bonded homodimer of ∼90 kDa type I transmembrane subunits
- CD105 is a component of the TGF-β receptor complex and is expressed by vascular endothelial smooth muscle cells, syncytiotrophoblasts of placenta and activated macrophages, and at relatively low levels by stromal fibroblasts
- Its expression is also observed in some types of tumors, and levels are up-regulated on the endothelium during angiogenesis
- CD105 has important roles in angiogenesis, cardiovascular development, and vascular remodeling, and the protein serves a regulatory role in cytoskeletal reorganization by modulating the sites of focal adhesion and cellular migration
- This antibody clone has been verified for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411) and MesenCult™-XF Medium (Catalog #05420)
Specifications
Specifications
| Antigen | CD105 (Endoglin) |
| Applications | Immunoprecipitation, Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 43A3 |
| Conjugate | Biotin |
| Host Species | Mouse |
| Quantity | 100 μg |
| Primary or Secondary | Primary |
| Target Species | Human, Mouse |
| Isotype | IgG1 κ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?