Learn More
CD105 (Endoglin) Monoclonal Antibody (MJ7/18), eFluor™ 450, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£181.00
Specifications
Antigen | CD105 (Endoglin) |
---|---|
Clone | MJ7/18 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15391740
|
Affymetrix eBioscience
48-1051-82 |
100 μg |
£181.00
100µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The MJ7/18 monoclonal antibody reacts with the mouse CD105 molecule, also known as Endoglin. This 90 kDa disulfide-linked homodimer is expressed by vascular endothelial cells. It is suggested that CD105 functions in adhesion and embryonic angiogenesis. Applications Reported: This MJ7/18 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MJ7/18 antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.Specifications
CD105 (Endoglin) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Q63961 | |
ENG | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
MJ7/18 | |
Flow Cytometry | |
eFluor 450 | |
Rat | |
Mouse | |
13805 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.