missing translation for 'onlineSavingsMsg'
Learn More

CD105 (Endoglin) Monoclonal Antibody (MJ7/18), APC, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-1051-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15527986

  • £167.00 / 100µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The MJ7/18 monoclonal antibody reacts with the mouse CD105 molecule, also known as Endoglin. This 90 kDa disulfide-linked homodimer is expressed by vascular endothelial cells. It is suggested that CD105 functions in adhesion and embryonic angiogenesis. Applications Reported: This MJ7/18 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MJ7/18 antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD105 (Endoglin) Monoclonal antibody specifically detects CD105 (Endoglin) in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD105 (Endoglin)
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q63961
ENG
Affinity chromatography
RUO
13805
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
MJ7/18
APC
ENG
AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein
Rat
100 μg
Primary
Mouse
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.