Learn More
CD103 Monoclonal Antibody (LF61), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD10304
Code : 50
Additional Details : Weight : 0.50000kg
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
CD103 Monoclonal antibody specifically detects CD103 in Human samples. It is validated for Flow Cytometry.Specifications
CD103 | |
Monoclonal | |
PE | |
Itgae | |
A530055J10; alpha-E1; alpha-M290; aM290; antigen CD103; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; CD103; DC; DCs; HML-1 antigen; human mucosal lymphocyte antigen 1, alpha polypeptide; HUMINAE; integrin alpha E; integrin alpha E, epithelial-associated; integrin alpha E1, epithelial-associated; integrin alpha M290; Integrin alpha-E; Integrin alpha-E heavy chain; Integrin alpha-E light chain; Integrin alpha-IEL; integrin subunit alpha E; integrin, alpha E; integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide); integrin, alpha E, epithelial-associated; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen | |
Mouse | |
purified | |
ASR | |
3682 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
LF61 | |
PBS with 10% sucrose, BSA and 0.1% sodium azide | |
P38570 | |
Itgae | |
Human CD103 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |