missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD103 Hamster anti-Mouse , PE, Clone: 20000000, BD Biosciences™
Hamster Monoclonal Antibody
Brand: BD Biosciences 566844
This item is not returnable.
View return policy
Description
CD103 Monoclonal antibody specifically detects CD103 in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD103 | |
| Monoclonal | |
| PE | |
| Itgae; integrin alpha-E; Integrin αIEL chain; αE; alpha-E1 | |
| Mouse intestinal intraepithelial lymphocytes | |
| RUO | |
| Mouse | |
| IgG2 κ |
| Flow Cytometry | |
| 20000000 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Hamster | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction