missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD101 Monoclonal Antibody (Moushi101), NovaFluor™ Yellow 730, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£154.00 - £297.00
Specifications
| Antigen | CD101 |
|---|---|
| Clone | Moushi101 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055639
|
Invitrogen™
M045T03Y07-A |
100 μg |
£297.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055212
|
Invitrogen™
M045T02Y07-A |
25 μg |
£154.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD101 Monoclonal antibody specifically detects CD101 in Mouse samples. It is validated for Flow CytometrySpecifications
| CD101 | |
| 0.1 mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| A8E0Y8 | |
| 630146 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| Moushi101 | |
| Flow Cytometry | |
| NovaFluor Yellow 730 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Cd101; CD101 antigen; CD101 molecule; cell surface glycoprotein V7; EWI immunoglobulin subfamily member EWI-101; EWI101; EWI-101; Glu-Trp-Ile EWI motif-containing protein 101; Gm1016; Gm734; IGSF2; Immunoglobulin superfamily member 2; immunoglobulin superfamily, member 2; leukocyte surface protein; RP11-27K13.2; V7 | |
| IgG2a κ | |
| Affinity chromatography | |
| CD101 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title