Learn More
Invitrogen™ CD100 (SEMA4D) Monoclonal Antibody (BMA12 (BMA-12)), Biotin, eBioscience™

Rat Monoclonal Antibody
Brand: Invitrogen™ 13-1001-82
Description
Description: The BMA12 monoclonal antibody reacts with mouse CD100, also known as Sema4D. Sema4D/CD100 is a 150kDa transmembrane protein expressed constitutively on T cells and is involved in the activation of B cells through its interaction with CD72. Interaction of these two proteins diminishes the negative signaling normally effected by CD72. CD100 is already expressed strongly on resting T cells and somewhat weakly on B cells, but activation upregulates expression on both cell types. Antigen expression has been reported in Spleen, thymus and lymph nodes. Applications Reported: The BMA12 (BMA-12) antibody has been reported for use in flow cytometric analysis. Applications Tested: The BMA12 (BMA-12) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
Semaphorins are neuronal chemorepellants that direct pioneering neurons during nervous system development. CD100 (also known as semaphorin 4d), a 150 kDa homodimer, is a novel semaphorin that induces B cells to aggregate and improves their viability in vitro. CD100 modifies CD40-CD40L B cell signaling by augmenting B cell aggregation and survival and down regulating CD23 expression. CD72 is a lymphocyte receptor for the class IV semaphorin CD100, which is involved in regulating B cell signaling. CD100 is expressed on resting and PHA stimulated T cells. The protein is weakly expressed on NK cells, EBV transformed B cells, monocytes and tumor/peripheral blood T cell lines, and at higher density on activated T cells and B cells.
Specifications
| CD100 (SEMA4D) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| O09126 | |
| Sema4d | |
| Affinity chromatography | |
| RUO | |
| 20354 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| BMA12 (BMA-12) | |
| Biotin | |
| Sema4d | |
| A8; BB18; C9orf164; CD100; coll-4; GR3; M-Sema G; M-sema-G; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4D; sema J; Sema4d; Semacl2; SEMAJ; semaphorin 4D; semaphorin H; semaphorin-4D; Semaphorin-C-like 2; semaphorin-J; Semcl2 | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.