missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD100 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587663
This item is not returnable.
View return policy
Description
Immunogen sequence: YNCYKGYLPR QCLKFRSALL IGKKKPKSDF CDREQSLKET LVEPGSFSQQ NGEHPKPALD TGYETEQDTI TSKVPTDRED SQRIDDLSAR DKPFDVKCEL KFADSDADGD; Positive Samples: 293T; Cellular Location: Cell membrane, Single-pass type I membrane protein.
Semaphorins are neuronal chemorepellants that direct pioneering neurons during nervous system development. CD100 (also known as semaphorin 4d), a 150 kDa homodimer, is a novel semaphorin that induces B cells to aggregate and improves their viability in vitro. CD100 modifies CD40-CD40L B cell signaling by augmenting B cell aggregation and survival and down regulating CD23 expression. CD72 is a lymphocyte receptor for the class IV semaphorin CD100, which is involved in regulating B cell signaling. CD100 is expressed on resting and PHA stimulated T cells. The protein is weakly expressed on NK cells, EBV transformed B cells, monocytes and tumor/peripheral blood T cell lines, and at higher density on activated T cells and B cells.
Specifications
| CD100 | |
| Polyclonal | |
| Unconjugated | |
| Sema4d | |
| A8; BB18; C9orf164; CD100; coll-4; GR3; M-Sema G; M-sema-G; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4D; sema J; Sema4d; Semacl2; SEMAJ; semaphorin 4D; semaphorin H; semaphorin-4D; Semaphorin-C-like 2; semaphorin-J; Semcl2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10507 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.79 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q92854 | |
| Sema4d | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 753-862 of human SEMA4D (NP_0063693). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction