missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD10 Mouse, APC, Clone: SN5c, enQuireBio™
Mouse Monoclonal Antibody
£136.00 - £1305.00
Specifications
Antigen | CD10 |
---|---|
Clone | SN5c |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961658
|
EnquireBio
QAB21-APC-25TESTS |
25 Tests |
£136.00
25 tests |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15971658
|
EnquireBio
QAB21-APC-500TESTS |
500 Tests |
£1305.00
500 tests |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD10 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
4311 | |
IgG1 κ | |
Protein A/G Purified |
SN5c | |
Flow Cytometry | |
APC | |
RUO | |
NP_009219 | |
MME | |
Primary | |
Store at 2-4°C. |