Learn More
CD10 Monoclonal Antibody (MEM-78), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen CD1005
Code : 50
Additional Details : Weight : 0.50000kg
Description
Allophycocyanin (APC) is a stable and highly soluble phycobiliprotein that provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing exceptional quantum yields and molar extinction coefficients.
CD10 Monoclonal antibody specifically detects CD10 in Human samples. It is validated for Flow Cytometry.Specifications
CD10 | |
Monoclonal | |
APC | |
Mme | |
6030454K05Rik; Atriopeptidase; C85356; CALLA; CD10; CMT2T; common acute lymphoblastic leukemia antigen; common acute lymphocytic leukemia antigen; Enkephalinase; EPN; membrane metallo endopeptidase; membrane metalloendopeptidase; membrane metallo-endopeptidase; membrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, CD10); membrane metallo-endopeptidase (neutral endopeptidase/enkephalinase); membrane metallo-endopeptidase variant 1; membrane metallo-endopeptidase variant 2; MME; NEP; neprilysin; neprilysin-390; neprilysin-411; neutral endopeptidase 24.11; SFE; Skin fibroblast elastase | |
Mouse | |
purified | |
ASR | |
4311 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
MEM-78 | |
PBS with BSA, sucrose and 0.1% sodium azide | |
P08473 | |
Mme | |
Human CD10 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |