missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD1.1 Monoclonal Antibody (CB3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528766
This item is not returnable.
View return policy
Description
CD1.1 Monoclonal Antibody for Flow
CD1.1 molecules, like MHC I and II, play an important role in the immune system by presenting lipid, glycolipid and lipopeptide antigen to T and NKT cells. The chicken CD1 gene is located in the chicken MHC B locus and has an important implication for the primordial MHC gene evolution. Two proteins have been identified - CD1.1 and CD1.2. Analysis of RNA from blood cells showed that both genes are expressed in Bu-1+ cells and CD8 alpha+ cells. CD1.2 is expressed in TCR gamma delta cells but is nearly undetectable in TCR alpha beta. The monoclonal antibody CB3 antibody can identify cells of the B cell lineage and recognizes CD1.1 but not CD1.2 on the surface of transfected mammalian cells.
Specifications
| CD1.1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| BBS with no preservative; pH 8.2 | |
| 0 | |
| CD1B | |
| Lymphocytes from the bursa of Fabricius of outbred chickens. | |
| 1 mL | |
| Primary | |
| Chicken | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| CB3 | |
| Unconjugated | |
| CD1B | |
| a polypeptide; CD1.1; CD1.2; CD1.2 antigen; CD1-2 antigen; CD1A; CD1A antigen, a polypeptide; CD1a molecule; CD1A2; CD1B; CD1b molecule; CD1c; chCD1-2; T-cell surface glycoprotein CD1A2 antigen; T-cell surface glycoprotein CD1b | |
| Mouse | |
| Purified | |
| RUO | |
| 425802 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction