missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1.1 antigen Antibody (CB3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28362
This item is not returnable.
View return policy
Description
CD1.1 antigen Monoclonal specifically detects CD1.1 antigen in Chicken samples. It is validated for Flow Cytometry.
Specifications
| CD1.1 antigen | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry <= 1 ug/10^6 cells | |
| CD1A | |
| Lymphocytes from the bursa of Fabricius of outbred chickens | |
| 0.1 mg | |
| Dendritic Cell Markers, Immunology | |
| 909 | |
| Chicken | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunocytochemistry | |
| CB3 | |
| Unconjugated | |
| CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Chicken CD1.1 | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction