missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CD EfficientFeed™ C AGT™ Nutrient Supplement

Product Code. 11320902
Change view
Click to view available options
Quantity:
1 L
10 L
100 L
Unit Size:
Each
Product Code. Quantity unitSize
11320902 1 L Each
10476359 10 L Each
11390892 100 L Each
3 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11320902

Brand: Gibco™ A1327504

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

CD EfficientFeed™ C AGT™ Nutrient Supplement

GIBCO™ CD EfficientFeed™ C AGT™ nutrient supplement is a dry format single part supplement for fed batch addition to the culture of multiple cell lines. This product is designed to work with GS CHO cell lines as a single feed without the need for additional GS supplements. CD EfficientFeed™ C AGT™ nutritient supplement is an animal origin-free, chemically defined formulation that contains no proteins, hydrolysates, or components of incompletely defined composition.

The AGT™ format is designed for large scale manufacturing and ease of use.
  • Up to 2.5 – 6 Fold increases versus batch culture – higher concentration of key components
  • Higher productivity in GS CHO and other selected cell lines
  • Avoid issues associated with the use of hydrolysates and other alternatives while gaining superior performance
  • Simplify scale-up by leveraging the value of AGT™ simple reconstitution
  • Ready for larger scale use with convenient packaging options (1 L, 10 L, and 100 L)
  • No components of animal origin, minimizing the opportunity for introduction of adventitious agents and facilitating the regulatory process

    Which EfficientFeed™ Nutrient Supplement to Choose?
    Consider CD EfficientFeed™ C AGT™ supplement when:
  • You want higher productivity in GS CHO and other selected cell lines
  • You are currently using CD FortiCHO™ as your base cell growth medium
  • You are using a base medium with greater than 6 g/L glucose or a hydrolysate at day 0

    Consider CHO CD EfficientFeed™ A supplement when:
  • You are currently using CD OptiCHO™ as your base cell growth medium

    Consider CHO CD EfficientFeed™ B supplement when:
  • You are currently using CD CHO as your base cell growth medium

    Consider the CHO CD Efficient Feed™ Kit when:
  • You are using a base medium with less than 6 g/L glucose and no hydrolysate at day 0

    Increased Titers
    CHO CD EfficientFeed™ supplements have been shown to fold increases in titers versus batch culture, and are an excellent starting point for process development. Furthermore, it does not require addition of a GS Supplement when used in CHO cell lines expressing endogenous glutamine synthetase.

    Simple Scale-Up and Cost Savings
    The ability to scale up in a dry format provides ease of handling and storage in manufacturing and other bioprocess settings. With AGT™ (Advanced Granulation Technology™) dry format, it is possible to store and use the product even more easily and safely than traditional dry format products. CD EfficientFeed™ C AGT™ supplement is offered in 1 L, 10 L and 100 L packaging that allows scale-up as a seamless process, as well as allow for cost savings when purchasing production level volumes.

    AGT™ Format Ease of Use
    CD EfficientFeed™ C AGT™ Nutrient Supplement delivers dry format scale-up convenience as single part AGT dry format. For more details, consult the supporting documents provided below.

    For research or further manufacturing use. Not for use in diagnostic procedures.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store in the dark at +2 to +8°C. Stable for 12 months when properly stored.
    Culture Environment CO2
    Antibiotic Spectrum No Antibiotics
    Cell Line CHO
    Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
    Culture Type Suspension Cell Culture
    Form AGT
    Product Type Media Supplement
    Serum Level Serum-Free
    Sterility Non-sterile
    With Additives Sodium Bicarbonate
    Without Additives No Glutamine, No Phenol Red, No Sodium Pyruvate, No Insulin
    Endotoxin Level Low
    Format Securitainer Capped Vial
    Purity or Quality Grade Cell Culture Grade, Qualified for Further Manufacturing, Research Grade
    Quantity 1 L
    Remarks Chemically Defined Nutrient Feeding Supplement
    Shipping Condition Wet Ice
    Source Animal origin-free
    Tested For Solubility
    Show More Show Less

    For Research Use or Further Manufacturing. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.