missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CD BHK-21 Production Medium

Product Code. p-7075673
Click to view available options
Quantity:
1 x 100 L
1 x 10 L
10 kg
Unit Size:
0.2g
100 L
10kg
This item is not returnable. View return policy

Product Code. 15349245

Brand: Gibco™ A1627701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CD BHK-21 Production Medium

Gibco™ CD BHK-21 Production Medium is a chemically defined, protein-free cell culture medium. It is designed to support the growth of BHK-21 cells in serum-free suspension culture. This medium is suitable for large-scale manufacturing applications.

CD BHK-21 Production Medium is manufactured as follows:

With

  • L-glutamine
  • Dry powder format

Without

  • Phenol Red
  • Protein hydrolysates

Gibco™ CD BHK-21 Production Medium is the first cell culture medium supporting complete serum-free and protein-free processes for BHK-21 cells. The dry powder format is easy to reconstitute, since it has no hydrolysates in the formulation. Adapting cells from serum-containing media is a rapid and simple procedure. BHK-21 cells can achieve high cell densities and maintain high cell viability in this medium.

Gibco™ CD BHK-21 Production Medium is suitable for large-scale suspension culture. Because it does not require supplementation with serum or other undefined components, Gibco™ CD BHK-21 Production Medium eliminates the risks associated with serum, while allowing you to achieve consistent virus titers and simplifying downstream purification.

cGMP Manufacturing
Gibco™ CD BHK-21 Production Medium is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line BHK-21
Form Powder
Product Type Production Medium
With Additives Glutamine
Without Additives No Phenol Red
For Use With (Application) Viral Vaccine Production
Quantity 1 x 10 L
Content And Storage Store in refrigerator (2–8°C).
Culture Type Suspension Cell Culture

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.