Learn More
Gibco™ CD BHK-21 Production Medium
Description
Gibco™ CD BHK-21 Production Medium is a chemically defined, protein-free cell culture medium. It is designed to support the growth of BHK-21 cells in serum-free suspension culture. This medium is suitable for large-scale manufacturing applications.
CD BHK-21 Production Medium is manufactured as follows:
With
- L-glutamine
- Dry powder format
Without
- Phenol Red
- Protein hydrolysates
Gibco™ CD BHK-21 Production Medium is the first cell culture medium supporting complete serum-free and protein-free processes for BHK-21 cells. The dry powder format is easy to reconstitute, since it has no hydrolysates in the formulation. Adapting cells from serum-containing media is a rapid and simple procedure. BHK-21 cells can achieve high cell densities and maintain high cell viability in this medium.
Gibco™ CD BHK-21 Production Medium is suitable for large-scale suspension culture. Because it does not require supplementation with serum or other undefined components, Gibco™ CD BHK-21 Production Medium eliminates the risks associated with serum, while allowing you to achieve consistent virus titers and simplifying downstream purification.
cGMP Manufacturing
Gibco™ CD BHK-21 Production Medium is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Specifications
| Cell Line | BHK-21 |
| Form | Powder |
| Product Type | Production Medium |
| With Additives | Glutamine, Phenol Red |
| Quantity | 1 x 100 L |
| Content And Storage | Store in refrigerator (2–8°C). |
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.