missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CD 3254

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective RXRα agonist; exhibits no activity at RARα, RARβ or RARγ receptors.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 364.48 |
| Purity | >97% |
| Molecular Weight (g/mol) | 364.485 |
| Chemical Name or Material | CD 3254 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction