missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CD 293 AGT™ Medium
Click to view available options
Quantity:
1 L
10 kg
10 L
100 L
Format:
Ropak Bucket
Securitainer Capped Vial
Unit Size:
1 L
10 L
100 L
10kg
Description
CD 293 AGT™ is a protein-free, chemically-defined medium in an easy-to-use granular format. CD 293 AGT™ is optimized for the growth of suspension cultures of 293 cells.
AGT = Advanced Granulation Technology
Specifications
Specifications
| Cell Line | 293 (HEK) |
| Classification | Animal Origin-free |
| Form | Powder (AGT) |
| Product Type | CD 293 Medium |
| Serum Level | Serum-free |
| Sterility | Non-sterile |
| With Additives | Low Glucose, Sodium Pyruvate, Sodium Bicarbonate, HEPES |
| Without Additives | No Glutamine |
| Glucose Concentration | 4500 mg/L |
| Quantity | 10 kg |
| Show More |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction