missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCZ1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | CCZ1 |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18698755
|
Novus Biologicals
NBP2-46711-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18624006
|
Novus Biologicals
NBP2-46711 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCZ1 Polyclonal antibody specifically detects CCZ1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| CCZ1 | |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| F5H553 | |
| 51622 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| C7orf28A, C7orf28B, CCZ1 vacuolar protein trafficking and biogenesis associated homolog (S. cerevisiae), CCZ1 vacuolar protein trafficking and biogenesis associated homolog B (S. cerevisiae), CCZ1A, CGI-43, chromosome 7 open reading frame 28B, FLJ60592, H_DJ1163J12.2, H_NH0577018.2, MGC19819, vacuolar fusion protein CCZ1 homolog-like | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: EMPGNLQHYGRFLTGPLNLNDPDAKCRFPKIFVNTDDTYEELHLIVYKAMSAAVCFMIDASVHPTLDFCRRLDSIVGPQLTVLASDICEQFNINKRMSGSEK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title