missing translation for 'onlineSavingsMsg'
Learn More

CCX354, MedChemExpress

Product Code. 16422568
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422568 5 mg 5mg
16410369 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16422568 Supplier Medchem Express Supplier No. HYU00350
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CCX354 is an antagonist of CCR1, with anti-inflammatory activity.

CCX354 is an antagonist of CCR1, with anti-inflammatory activity.

Specifications

Chemical Name or Material CCX354
CAS 1010073-75-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H22ClN7O2
Quantity 5 mg
SMILES COC1=CC(N2CCN(C(CN3N=C(C4=NC=CN4)C5=CC=CN=C53)=O)CC2)=CC=C1Cl
Molecular Weight (g/mol) 451.91
Formula Weight 451.91
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.