missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT6B Polyclonal Antibody

CCT6B Polyclonal Antibody
Brand: Invitrogen PA5100432
This item is not returnable.
View return policy
Description
CCT6B Polyclonal Antibody for Western Blot
Specifications
| CCT6B | |
| Polyclonal | |
| Unconjugated | |
| Cct6b | |
| Cct6b; Cctz2; Cctz-2; CCT-zeta-2; CCTzeta-2; CCT-zeta-like; chaperonin containing TCP1 subunit 6B; chaperonin containing TCP1, subunit 6B (zeta 2); chaperonin containing Tcp1, subunit 6b (zeta); chaperonin subunit 6b (zeta); chaperonin-containing T-complex polypeptide 1, subunit 6B; T-complex protein 1 subunit zeta-2; TCP-1-zeta-2; TCP-1-zeta-like; Testis-specific protein TSA303; testis-specific Tcp20; TSA303 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10693, 12467, 363658 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q61390, Q92526 | |
| Cct6b | |
| A synthesized peptide derived from human CCT6B(Accession Q92526), corresponding to amino acid residues I275-S325. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction