missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT6B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92177-0.1ml
This item is not returnable.
View return policy
Description
CCT6B Polyclonal antibody specifically detects CCT6B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| CCT6B | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Cctz2, CCTZ-2, CCT-zeta-2, CCT-zeta-like, chaperonin containing TCP1, subunit 6B (zeta 2), T-complex protein 1 subunit zeta-2, T-complex protein 1, zeta-2 subunit, TCP-1-zeta-2, TCP-1-zeta-like, Testis-specific protein TSA303, Testis-specific Tcp20, TSA303 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-340 of human CCT6B (NP_006575.2). AEGFEAAKIKALEVLEEVKVTKEMKRKILLDVARTSLQTKVHAELADVLTEVVVDSVLAVRRPGYPIDLFMVEIMEMKHKLGTDTKLIQGLVLDHGARHPDMKKRVEDAFILICNVSLEYEKTEVNSGFFYKTAEEKEKLVKAERKFIEDRVQKIIDLKDKVCAQSNKGFVVINQKGIDPFSLDSLAKHGIVALRRAKRRNMERLSLACGGMAVNSFEDLT | |
| 0.1 mL | |
| Signal Transduction | |
| 10693 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction