missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT6A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92778-0.1ml
This item is not returnable.
View return policy
Description
CCT6A Polyclonal antibody specifically detects CCT6A in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CCT6A | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| acute morphine dependence related protein 2, Acute morphine dependence-related protein 2, amino acid transport defect-complementing, CCT6, Cctz, CCT-zeta, CCT-zeta-1, chaperonin containing T-complex subunit 6, chaperonin containing TCP1, subunit 6A (zeta 1), histidine transport regulator 3, HTR3MGC126215, MGC126214, MoDP-2, T-complex protein 1 subunit zeta, T-complex protein 1, zeta subunit, TCP-1-zeta, TCP20, TCPZ, TTCP20 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 80-250 of human CCT6A (NP_001753.1). VATAQDDITGDGTTSNVLIIGELLKQADLYISEGLHPRIITEGFEAAKEKALQFLEEVKVSREMDRETLIDVARTSLRTKVHAELADVLTEAVVDSILAIKKQDEPIDLFMIEIMEMKHKSETDTSLIRGLVLDHGARHPDMKKRVEDAYILTCNVSLEYEKTEVNSGFFY | |
| 0.1 mL | |
| Signal Transduction | |
| 908 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction