Learn More
CCT5 Mouse anti-Human, Unconjugated, Clone: 1B12E10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67400-1-IG-20UL
 View more versions of this product
Product Code. 16870006
  • £135.00 / 20µL
Description
The CCT5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCT5. This antibody recognizes human, mouse, rat antigen. The CCT5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
CCT5
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
1B12E10
Unconjugated
20 μL
22948
IgG1