Learn More
CCT5 Mouse anti-Human, Unconjugated, Clone: 1B12E10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67400-1-IG-150UL
This item is not returnable.
View return policy
Description
The CCT5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCT5. This antibody recognizes human, mouse, rat antigen. The CCT5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CCT5 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, ELISA | |
| 1B12E10 | |
| Unconjugated | |
| 150 μL | |
| 22948 | |
| IgG1 |