missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10571-1-AP-20UL
This item is not returnable.
View return policy
Description
CCT3 (Chaperonin Containing TCP1 Subunit 3) is a molecular chaperone that is a member of the chaperonin containing TCP1 complex (CCT). The complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants have been characterized for this gene. In addition, a pseudogene of this gene has been found on chromosome 8. Diseases associated with CCT3 include Spinocerebellar Ataxia 10 and Immature Cataract.Specifications
| CCT3 | |
| Polyclonal | |
| Unconjugated | |
| CCT3 | |
| CCT gamma, CCT3, TCP 1 gamma | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12462, 295230, 7203 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P49368, P80318, Q6P502 | |
| CCT3 | |
| CCT3 Fusion Protein Ag0843 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction