missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT3 Mouse anti-Human, Mouse, Clone: 5F5A6, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CCT3 |
|---|---|
| Clone | 5F5A6 |
| Concentration | 0.85 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16867315
|
Proteintech
60264-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16857315
|
Proteintech
60264-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a molecular chaperone that is a member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants have been characterized for this gene. In addition, a pseudogene of this gene has been found on chromosome 8.Specifications
| CCT3 | |
| 0.85 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CCT gamma, CCT3, CCTG, hTRiC5, PIG48, TCP 1 gamma, TRIC5 | |
| CCT3 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 5F5A6 | |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Mouse, Human | |
| P49368, P80318 | |
| 12462, 7203 | |
| CCT3 Fusion Protein Ag0843 | |
| Primary | |
| -20°C | |
| CCT3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title