missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CCSER2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA557968

Product Code. 15725861

  • £506.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: NVECDNMNRF DRPDRNVRQP QEGFWKRPPQ RWSGQEHYHL SHPDHYHHHG KSDLSRGSPY RESPLGHFES YGGMPFFQAQ KMFVDVPENT VILDEMT Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CCSER2
Polyclonal
Unconjugated
CCSER2
1700012P13Rik; 2900054P12Rik; AI035535; AW108503; bA486O22.1; Ccser2; coiled-coil serine rich 2; coiled-coil serine rich protein 2; coiled-coil serine-rich protein 2; FAM190B; family with sequence similarity 190, member B; Gcap14; granule cell antiserum positive 14; granule cell antiserum positive protein 14; KIAA1128; NPD012; protein FAM190B; Protein GCAP14; Protein GCAP14 homolog; RGD1304626; serine-rich coiled-coil domain-containing protein 2
Rabbit
Antigen affinity chromatography
RUO
306306, 54462, 72972
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.8 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q3UHI0, Q9H7U1
CCSER2
Recombinant protein corresponding to Human CCSER2. Recombinant protein control fragment (Product #RP-97047).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.