missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCS/SOD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£513.00
Specifications
| Antigen | CCS/SOD4 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18261055
|
Novus Biologicals
NBP2-55196 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677486
|
Novus Biologicals
NBP2-55196-25ul |
25 μL |
N/A
|
N/A | |||||
Description
CCS/SOD4 Polyclonal specifically detects CCS/SOD4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CCS/SOD4 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| copper chaperone for superoxide dismutase, MGC138260, Superoxide dismutase copper chaperone | |
| CCS | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Cancer | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 9973 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SDSGNQGTLCTLEFAVQMTCQSCVDAVRKSLQGVAGVQDVEVHLEDQMVLVHTTLPSQEVQALLEGTGRQAVLKGMGS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title